Assay Details
Gene Symbol: PTK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688276.3 | XP_005688333.2 | 4 - 5 | 781 | 69 | |
GenBank mRNA | KA329840.1 | - | 4 - 5 | 974 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
