Assay Details
| Gene Symbol: LEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285746.1 | NP_001272675.1 | 1 - 2 | 212 | 58 | |
| XM_005681326.3 | XP_005681383.1 | 1 - 2 | 1394 | 58 | ||
| XM_018049144.1 | XP_017904633.1 | 1 - 2 | 703 | 58 | ||
| XM_018049145.1 | XP_017904634.1 | 1 - 2 | 1394 | 58 | ||
| GenBank mRNA | HM059927.1 | - | 1 - 2 | 212 | 58 | |
More Information
Set Membership: |
|

LEF1
Genomic Map