Assay Details
| Gene Symbol: LOC102171917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065326.1 | XP_017920815.1 | 23 - 24 | 3501 | 77 | |
| XM_018065327.1 | XP_017920816.1 | 12 - 13 | 2135 | 77 | ||
More Information
Set Membership: |
|

LOC102171917
Genomic Map