Assay Details
| Gene Symbol: TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046181.1 | XP_017901670.1 | 4 - 5 | 598 | 74 | |
| GenBank mRNA | JO415931.2 | - | 1 - 2 | 138 | 74 | |
| JO451401.2 | - | 1 - 2 | 139 | 74 | ||
| JO585703.2 | - | 4 - 5 | 546 | 74 | ||
| JO638327.2 | - | 4 - 5 | 559 | 74 | ||
| JT705887.1 | - | 1 - 2 | 65 | 74 | ||
More Information
Set Membership: |
|

TCTEX1D2
Genomic Map