Assay Details
| Gene Symbol: LOC102184834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060680.1 | XP_017916169.1 | 11 - 12 | 1369 | 80 | |
| XM_018060681.1 | XP_017916170.1 | 11 - 12 | 1369 | 80 | ||
| XM_018060682.1 | XP_017916171.1 | 12 - 13 | 2673 | 80 | ||
| XM_018060683.1 | XP_017916172.1 | 12 - 13 | 2673 | 80 | ||
| XM_018060684.1 | XP_017916173.1 | 12 - 13 | 2673 | 80 | ||
More Information
Set Membership: |
|

LOC102184834
Genomic Map