Assay Details
| Gene Symbol: LOC102184834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060680.1 | XP_017916169.1 | 17 - 18 | 1967 | 75 | |
| XM_018060682.1 | XP_017916171.1 | 18 - 19 | 3271 | 75 | ||
| XM_018060683.1 | XP_017916172.1 | 18 - 19 | 3271 | 75 | ||
More Information
Set Membership: |
|

LOC102184834
Genomic Map