Assay Details
| Gene Symbol: LOC102184834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060682.1 | XP_017916171.1 | 1 - 2 | 1298 | 79 | |
| XM_018060683.1 | XP_017916172.1 | 1 - 2 | 1298 | 79 | ||
| XM_018060684.1 | XP_017916173.1 | 1 - 2 | 1298 | 79 | ||
| GenBank mRNA | JO648925.2 | - | 2 - 3 | 266 | 79 | |
More Information
Set Membership: |
|

LOC102184834
Genomic Map