Assay Details
| Gene Symbol: LOC102184834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060680.1 | XP_017916169.1 | 23 - 24 | 3351 | 63 | |
| XM_018060681.1 | XP_017916170.1 | 22 - 23 | 3294 | 63 | ||
| XM_018060682.1 | XP_017916171.1 | 24 - 25 | 4655 | 63 | ||
| XM_018060683.1 | XP_017916172.1 | 24 - 25 | 4655 | 63 | ||
| XM_018060684.1 | XP_017916173.1 | 23 - 24 | 4598 | 63 | ||
More Information
Set Membership: |
|

LOC102184834
Genomic Map