Assay Details
| Gene Symbol: VGLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054393.1 | XP_017909882.1 | 2 - 3 | 767 | 88 | |
| XM_018054438.1 | XP_017909927.1 | 2 - 3 | 766 | 88 | ||
| XM_018054497.1 | XP_017909986.1 | 2 - 3 | 587 | 88 | ||
| XM_018054543.1 | XP_017910032.1 | 2 - 3 | 595 | 88 | ||
| GenBank mRNA | KA316917.1 | - | 1 - 2 | 204 | 88 | |
More Information
Set Membership: |
|

VGLL3
Genomic Map