Assay Details
| Gene Symbol: PCDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061533.1 | XP_017917022.1 | 3 - 4 | 3847 | 74 | |
| GenBank mRNA | JT722943.1 | - | 2 - 3 | 908 | 74 | |
More Information
Set Membership: |
|

PCDH10
Genomic Map