Assay Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969841.2 | XP_013825295.2 | 4 - 5 | 1075 | 68 | |
| XM_018062636.1 | XP_017918125.1 | 3 - 4 | 1070 | 68 | ||
More Information
Set Membership: |
|

MBD5
Genomic Map