Assay Details
| Gene Symbol: EMSY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690007.3 | XP_005690064.1 | 19 - 20 | 3154 | 62 | |
| XM_005690008.3 | XP_005690065.1 | 18 - 19 | 3111 | 62 | ||
| XM_013969807.2 | XP_013825261.1 | 18 - 19 | 3112 | 62 | ||
| XM_013969808.2 | XP_013825262.1 | 18 - 19 | 3001 | 62 | ||
| XM_013969809.2 | XP_013825263.1 | 18 - 19 | 2990 | 62 | ||
| XM_013969810.2 | XP_013825264.1 | 17 - 18 | 2958 | 62 | ||
| XM_013969811.2 | XP_013825265.1 | 16 - 17 | 2790 | 62 | ||
| XM_013969812.2 | XP_013825266.1 | 19 - 20 | 3155 | 62 | ||
| XM_018059320.1 | XP_017914809.1 | 18 - 19 | 3054 | 62 | ||
More Information
Set Membership: |
|

EMSY
Genomic Map