Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678404.3 | XP_005678461.2 | 5 - 6 | 1366 | 85 | |
| XM_018044181.1 | XP_017899670.1 | 6 - 7 | 1505 | 85 | ||
| XM_018044190.1 | XP_017899679.1 | 5 - 6 | 1485 | 85 | ||
More Information
Set Membership: |
|

TUT4
Genomic Map