Assay Details
| Gene Symbol: AOPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052467.1 | XP_017907956.1 | 15 - 16 | 2527 | 60 | |
| XM_018052468.1 | XP_017907957.1 | 12 - 13 | 2209 | 60 | ||
More Information
Set Membership: |
|

AOPEP
Genomic Map