Assay Details
| Gene Symbol: LOC102173333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045796.1 | XP_017901285.1 | 16 - 17 | 3175 | 68 | |
| XM_018045808.1 | XP_017901297.1 | 16 - 17 | 3136 | 68 | ||
| XM_018045817.1 | XP_017901306.1 | 16 - 17 | 3175 | 68 | ||
More Information
Set Membership: |
|

LOC102173333
Genomic Map