Assay Details
| Gene Symbol: LOC102173333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045796.1 | XP_017901285.1 | 27 - 28 | 4539 | 87 | |
| XM_018045804.1 | XP_017901293.1 | 27 - 28 | 4512 | 87 | ||
| XM_018045808.1 | XP_017901297.1 | 27 - 28 | 4500 | 87 | ||
| XM_018045812.1 | XP_017901301.1 | 27 - 28 | 4473 | 87 | ||
| XM_018045817.1 | XP_017901306.1 | 27 - 28 | 4539 | 87 | ||
| GenBank mRNA | KA337271.1 | - | 3 - 4 | 244 | 87 | |
More Information
Set Membership: |
|

LOC102173333
Genomic Map