Assay Details
Gene Symbol: LOC108635581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045824.1 | XP_017901313.1 | 23 - 24 | 3180 | 111 | |
XM_018045830.1 | XP_017901319.1 | 23 - 24 | 3178 | 111 | ||
XM_018045834.1 | XP_017901323.1 | 24 - 25 | 3281 | 111 |
Gene Symbol: LOC102173333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO444814.2 | - | 5 - 6 | 652 | 111 | |
JT717549.1 | - | 5 - 6 | 652 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
