Assay Details
| Gene Symbol: CITED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044038.1 | XP_017899527.1 | 2 - 3 | 719 | 84 | |
| XM_018044039.1 | XP_017899528.1 | 2 - 3 | 282 | 84 | ||
| XM_018044040.1 | XP_017899529.1 | 3 - 4 | 219 | 84 | ||
| XM_018044041.1 | XP_017899530.1 | 3 - 4 | 215 | 84 | ||
More Information
Set Membership: |
|

CITED1
Genomic Map