Assay Details
| Gene Symbol: IGFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285763.1 | NP_001272692.1 | 3 - 4 | 663 | 93 | |
| GenBank mRNA | HM185495.1 | - | 3 - 4 | 663 | 93 | |
More Information
Set Membership: |
|

IGFBP1
Genomic Map