Assay Details
| Gene Symbol: PELI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043221.1 | XP_017898710.1 | 6 - 7 | 847 | 84 | |
| XM_018043222.1 | XP_017898711.1 | 5 - 6 | 772 | 84 | ||
| GenBank mRNA | JO621225.2 | - | 5 - 6 | 692 | 84 | |
More Information
Set Membership: |
|

PELI3
Genomic Map