Assay Details
| Gene Symbol: POLR2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675184.3 | XP_005675241.1 | 1 - 2 | 1259 | 84 | |
| GenBank mRNA | JO442918.2 | - | 1 - 2 | 324 | 84 | |
| JO510089.2 | - | 4 - 5 | 627 | 84 | ||
More Information
Set Membership: |
|

POLR2H
Genomic Map