Assay Details
| Gene Symbol: SOX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693521.3 | XP_005693578.2 | 1 - 2 | 774 | 62 | |
| GenBank mRNA | JT711586.1 | - | 12 - 13 | 2422 | 62 | |
More Information
Set Membership: |
|

SOX15
Genomic Map