Assay Details
| Gene Symbol: STRADB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060886.1 | XP_017916375.1 | 9 - 10 | 1118 | 64 | |
| XM_018060888.1 | XP_017916377.1 | 9 - 10 | 991 | 64 | ||
| GenBank mRNA | JI861882.1 | - | 7 - 8 | 820 | 64 | |
| JO402874.2 | - | 8 - 9 | 904 | 64 | ||
| JO429104.2 | - | 3 - 4 | 1109 | 64 | ||
| JT748535.1 | - | 8 - 9 | 859 | 64 | ||
More Information
Set Membership: |
|

STRADB
Genomic Map