Assay Details
| Gene Symbol: FSD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052548.1 | XP_017908037.1 | 13 - 14 | 1553 | 74 | |
| XM_018052549.1 | XP_017908038.1 | 12 - 13 | 1520 | 74 | ||
| XM_018052550.1 | XP_017908039.1 | 12 - 13 | 1517 | 74 | ||
More Information
Set Membership: |
|

FSD1L
Genomic Map