Assay Details
| Gene Symbol: RND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065162.1 | XP_017920651.1 | 2 - 3 | 233 | 71 | |
| XM_018065163.1 | XP_017920652.1 | 2 - 3 | 289 | 71 | ||
| GenBank mRNA | JO634618.2 | - | 2 - 3 | 344 | 71 | |
| KA339441.1 | - | 2 - 3 | 324 | 71 | ||
More Information
Set Membership: |
|

RND2
Genomic Map