Assay Details
| Gene Symbol: MRTFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974838.2 | XP_013830292.2 | 3 - 4 | 875 | 76 | |
| XM_018040528.1 | XP_017896017.1 | 6 - 7 | 547 | 76 | ||
| XM_018040529.1 | XP_017896018.1 | 6 - 7 | 547 | 76 | ||
| XM_018040530.1 | XP_017896019.1 | 5 - 6 | 484 | 76 | ||
| XM_018040531.1 | XP_017896020.1 | 5 - 6 | 484 | 76 | ||
| XM_018040532.1 | XP_017896021.1 | 3 - 4 | 882 | 76 | ||
| XM_018040534.1 | XP_017896023.1 | 6 - 7 | 547 | 76 | ||
| XM_018040535.1 | XP_017896024.1 | 5 - 6 | 484 | 76 | ||
| GenBank mRNA | JO438317.2 | - | 4 - 5 | 424 | 76 | |
| KA347829.1 | - | 478 | 76 | |||
More Information
Set Membership: |
|

MRTFB
Genomic Map