Assay Details
| Gene Symbol: LRRC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062145.1 | XP_017917634.1 | 7 - 8 | 1380 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC36| Gene Symbol: LRRC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062145.1 | XP_017917634.1 | 7 - 8 | 1380 | 63 | |
Set Membership: |
|