Assay Details
| Gene Symbol: ZMPSTE24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678642.3 | XP_005678699.1 | 5 - 6 | 844 | 63 | |
| XM_018042306.1 | XP_017897795.1 | 4 - 5 | 628 | 63 | ||
| GenBank mRNA | JO585628.2 | - | 4 - 5 | 738 | 63 | |
More Information
Set Membership: |
|

ZMPSTE24
Genomic Map