Assay Details
| Gene Symbol: RGS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690464.3 | XP_005690521.2 | 3 - 4 | 467 | 113 | |
| XM_018060153.1 | XP_017915642.1 | 2 - 3 | 365 | 113 | ||
| GenBank mRNA | JO576111.1 | - | 1 - 2 | 168 | 113 | |
More Information
Set Membership: |
|

RGS18
Genomic Map