Assay Details
Gene Symbol: LOC102176695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054881.1 | XP_017910370.1 | 10 - 11 | 1653 | 101 | |
XM_018054882.1 | XP_017910371.1 | 11 - 12 | 1719 | 101 | ||
GenBank mRNA | JO339437.2 | - | 10 - 11 | 2719 | 101 | |
JO580143.1 | - | 8 - 9 | 2301 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
