Assay Details
| Gene Symbol: LOC102172279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058445.1 | XP_017913934.1 | 13 - 14 | 1216 | 85 | |
| XM_018058446.1 | XP_017913935.1 | 13 - 14 | 1201 | 85 | ||
| XM_018058447.1 | XP_017913936.1 | 12 - 13 | 1132 | 85 | ||
More Information
Set Membership: |
|

LOC102172279
Genomic Map