Assay Details
| Gene Symbol: WWTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049076.1 | XP_017904565.1 | 3 - 4 | 859 | 57 | |
| XM_018049081.1 | XP_017904570.1 | 3 - 4 | 863 | 57 | ||
| GenBank mRNA | JO425860.1 | - | 4 - 5 | 2659 | 57 | |
More Information
Set Membership: |
|

WWTR1
Genomic Map