Assay Details
| Gene Symbol: RASSF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679780.3 | XP_005679837.1 | 1 - 2 | 418 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RASSF9| Gene Symbol: RASSF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679780.3 | XP_005679837.1 | 1 - 2 | 418 | 95 | |
Set Membership: |
|