Assay Details
| Gene Symbol: FGF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685797.3 | XP_005685854.2 | 2 - 3 | 857 | 86 | |
| GenBank mRNA | JT706907.1 | - | 3 - 4 | 435 | 86 | |
More Information
Set Membership: |
|

FGF7
Genomic Map