Assay Details
| Gene Symbol: PPP1R3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700737.3 | XP_005700794.3 | 2 - 3 | 4128 | 75 | |
| XM_018045138.1 | XP_017900627.1 | 2 - 3 | 4053 | 75 | ||
| GenBank mRNA | JO639149.2 | - | 2 - 3 | 1833 | 75 | |
| JT705173.1 | - | 9 - 10 | 1483 | 75 | ||
More Information
Set Membership: |
|

PPP1R3F
Genomic Map