Assay Details
| Gene Symbol: EPHX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051295.1 | XP_017906784.1 | 5 - 6 | 1665 | 56 | |
| XM_018051296.1 | XP_017906785.1 | 6 - 7 | 821 | 56 | ||
| XM_018051297.1 | XP_017906786.1 | 5 - 6 | 1665 | 56 | ||
| XM_018051298.1 | XP_017906787.1 | 5 - 6 | 1665 | 56 | ||
| GenBank mRNA | JO617569.2 | - | 5 - 6 | 1724 | 56 | |
More Information
Set Membership: |
|

EPHX3
Genomic Map