Assay Details
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059714.1 | XP_017915203.1 | 3 - 4 | 594 | 69 | |
GenBank mRNA | JO425699.1 | - | 7 - 8 | 3008 | 69 | |
JO425709.1 | - | 7 - 8 | 3002 | 69 | ||
JO496249.2 | - | 3 - 4 | 606 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
