Assay Details
| Gene Symbol: LOC108633221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044427.1 | XP_017899916.1 | 130 | 97 | ||
| GenBank mRNA | JO367983.2 | - | 551 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018044427.1 transcripts.
Set Membership: |
|

LOC108633221
Genomic Map