Assay Details
| Gene Symbol: LOC108633221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044427.1 | XP_017899916.1 | 1 - 2 | 253 | 69 | |
| GenBank mRNA | JO367983.2 | - | 4 - 5 | 428 | 69 | |
More Information
Set Membership: |
|

LOC108633221
Genomic Map