Assay Details
| Gene Symbol: RAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043455.1 | XP_017898944.1 | 4 - 5 | 398 | 66 | |
| GenBank mRNA | JO643558.2 | - | 4 - 5 | 405 | 66 | |
More Information
Set Membership: |
|

RAB1B
Genomic Map