Assay Details
| Gene Symbol: HOMER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685075.3 | XP_005685132.1 | 8 - 9 | 1964 | 66 | |
| GenBank mRNA | JT716212.1 | - | 3 - 4 | 607 | 66 | |
More Information
Set Membership: |
|

HOMER1
Genomic Map