Assay Details
| Gene Symbol: SUMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063832.1 | XP_017919321.1 | 2 - 3 | 312 | 107 | |
| GenBank mRNA | JO506347.2 | - | 2 - 3 | 701 | 107 | |
More Information
Set Membership: |
|

SUMO2
Genomic Map