Assay Details
| Gene Symbol: SEPT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063712.1 | XP_017919201.1 | 9 - 10 | 1557 | 54 | |
| XM_018063713.1 | XP_017919202.1 | 8 - 9 | 2299 | 54 | ||
| XM_018063714.1 | XP_017919203.1 | 8 - 9 | 1646 | 54 | ||
| XM_018063715.1 | XP_017919204.1 | 7 - 8 | 920 | 54 | ||
| XM_018063716.1 | XP_017919205.1 | 7 - 8 | 812 | 54 | ||
| XM_018063717.1 | XP_017919206.1 | 7 - 8 | 774 | 54 | ||
| XM_018063718.1 | XP_017919207.1 | 7 - 8 | 907 | 54 | ||
More Information
Set Membership: |
|

SEPT9
Genomic Map