Assay Details
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285627.1 | NP_001272556.1 | 2 - 3 | 634 | 55 | |
XM_018039333.1 | XP_017894822.1 | 3 - 4 | 915 | 55 | ||
XM_018039334.1 | XP_017894823.1 | 3 - 4 | 824 | 55 | ||
XM_018039335.1 | XP_017894824.1 | 3 - 4 | 637 | 55 | ||
GenBank mRNA | EF486267.1 | - | 2 - 3 | 634 | 55 | |
KA317434.1 | - | 3 - 4 | 625 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
