Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285662.1 | NP_001272591.1 | 4 - 5 | 335 | 62 | |
| XM_013965444.2 | XP_013820898.1 | 5 - 6 | 475 | 62 | ||
| GenBank mRNA | EU239656.1 | - | 4 - 5 | 335 | 62 | |
More Information
Set Membership: |
|

SPARC
Genomic Map