Assay Details
| Gene Symbol: PNPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285739.1 | NP_001272668.1 | 4 - 5 | 837 | 64 | |
| XM_018042656.1 | XP_017898145.1 | 5 - 6 | 917 | 64 | ||
| XM_018042657.1 | XP_017898146.1 | 4 - 5 | 1266 | 64 | ||
| GenBank mRNA | GQ918145.1 | - | 4 - 5 | 837 | 64 | |
More Information
Set Membership: |
|

PNPLA2
Genomic Map