Assay Details
| Gene Symbol: CYP19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285747.1 | NP_001272676.1 | 4 - 5 | 662 | 85 | |
| XM_013967046.2 | XP_013822500.2 | 5 - 6 | 855 | 85 | ||
| GenBank mRNA | AY148883.1 | - | 4 - 5 | 662 | 85 | |
More Information
Set Membership: |
|

CYP19A1
Genomic Map