Assay Details
| Gene Symbol: IL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285758.1 | NP_001272687.1 | 3 - 4 | 463 | 69 | |
| GenBank mRNA | HM542482.1 | - | 3 - 4 | 463 | 69 | |
More Information
Set Membership: |
|

IL22
Genomic Map