Assay Details
| Gene Symbol: CTSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314245.1 | NP_001301174.1 | 6 - 7 | 705 | 80 | |
| XM_018051778.1 | XP_017907267.1 | 7 - 8 | 840 | 80 | ||
| XM_018051779.1 | XP_017907268.1 | 3 - 4 | 305 | 80 | ||
| GenBank mRNA | JN986831.1 | - | 6 - 7 | 705 | 80 | |
| JO359571.2 | - | 1466 | 80 | |||
More Information
Set Membership: |
|

CTSB
Genomic Map