Assay Details
| Gene Symbol: TNNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285572.1 | NP_001272501.1 | 4 - 5 | 338 | 62 | |
| GenBank mRNA | HQ640744.1 | - | 4 - 5 | 338 | 62 | |
More Information
Set Membership: |
|

TNNC1
Genomic Map